Assay Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018046.4 | NP_060516.2 | 1 - 2 | 595 | 80 | |
| GenBank mRNA | AW958325.1 | - | 1 - 2 | 77 | 80 | |
| AY500994.1 | - | 1 - 2 | 514 | 80 | ||
| BC032844.1 | - | 1 - 2 | 581 | 5266978 | 80 | |
| BI561566.1 | - | 1 - 2 | 599 | 80 | ||
| U84971.1 | - | 1 - 2 | 524 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000377.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05005267.hg.1 | TC05000377.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005267.hg.1 | ENST00000312916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005267.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005267.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009640.hg.1 | ENST00000509500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009478.hg.1 | ENST00000426656.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005264.hg.1 | TC05000377.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005264.hg.1 | ENST00000312916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005264.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005265.hg.1 | TC05000377.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005265.hg.1 | ENST00000312916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005265.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005265.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005268.hg.1 | TC05000377.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005268.hg.1 | ENST00000312916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005268.hg.1 | ENST00000503538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005268.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005268.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005266.hg.1 | TC05000377.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005266.hg.1 | ENST00000312916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005266.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005266.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153664.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153664.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153664.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153664.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153664.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153664.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153664.hg.1 | TC0500007841.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500153663.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153663.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153663.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153663.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153663.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153663.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153663.hg.1 | TC0500007841.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153661.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153661.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153661.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153661.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153661.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153661.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153661.hg.1 | TC0500007841.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000009478.hg.1 | ENST00000426656.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153662.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153662.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153662.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153662.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153662.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153662.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153662.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153662.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153662.hg.1 | TC0500007841.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009640.hg.1 | ENST00000509500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153665.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153665.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153665.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153665.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153665.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153665.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153665.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153665.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153665.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153665.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153665.hg.1 | TC0500007841.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 117353 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 224660 | - | - |

AGGF1
Genomic Map