Assay Details
| Gene Symbol: TMEM50A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014313.3 | NP_055128.1 | 1 - 2 | 161 | 71 | |
| XM_005245817.1 | XP_005245874.1 | 1 - 2 | 125 | 71 | ||
| GenBank mRNA | AF081282.1 | - | 1 - 2 | 88 | 71 | |
| AF087880.1 | - | 1 - 2 | 149 | 71 | ||
| AK299180.1 | - | 1 - 2 | 125 | 71 | ||
| AK312588.1 | - | 1 - 2 | 125 | 71 | ||
| AL136627.1 | - | 1 - 2 | 143 | 71 | ||
| AY071927.1 | - | 1 - 2 | 138 | 71 | ||
| AY358650.1 | - | 1 - 2 | 124 | 71 | ||
| BC007341.2 | - | 1 - 2 | 109 | 3688672 | 71 | |
| BC066908.1 | - | 1 - 2 | 100 | 4817784 | 71 | |
| JX644905.1 | - | 1 - 2 | 161 | 71 | ||
| JX644906.1 | - | 1 - 2 | 161 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000319.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF081282.1,AF087880.1,AK299180.1,AK312588.1,AL136627.1,AY071927.1,AY358650.1,BC007341.2,BC066908.1,JX644905.1,JX644906.1,NM_014313.3,XM_005245817.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01004889.hg.1 | TC01000319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004889.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004889.hg.1 | ENST00000480937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004889.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004891.hg.1 | TC01000319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004891.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004891.hg.1 | ENST00000480937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004891.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004887.hg.1 | TC01000319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004887.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004887.hg.1 | ENST00000480937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004887.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000487234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000487234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000487234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151538.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151538.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151538.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151538.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151538.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151538.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151538.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151538.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151538.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151538.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151538.hg.1 | TC0100007398.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000487234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000487234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000487234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007398.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151541.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151541.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151541.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151541.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151541.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151541.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151541.hg.1 | ENST00000468704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151541.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151541.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151541.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151541.hg.1 | TC0100007398.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100151536.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151536.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151536.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151536.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151536.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151536.hg.1 | ENST00000374358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151536.hg.1 | ENST00000480937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151536.hg.1 | ENST00000491936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151536.hg.1 | ENST00000498135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151536.hg.1 | NM_014313.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151536.hg.1 | TC0100007398.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 207955 | - | - |

TMEM50A
Genomic Map