Assay Details
| Gene Symbol: PACSIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF130979.1 | - | 1 - 2 | 112 | 142 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF130979.1 transcripts.
Set Membership: |
|

PACSIN3
Genomic Map