Assay Details
| Gene Symbol: GNG13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016541.2 | NP_057625.1 | 1 - 2 | 74 | 77 | |
| GenBank mRNA | AB030207.1 | - | 1 - 2 | 53 | 77 | |
| AK312139.1 | - | 1 - 2 | 75 | 77 | ||
| AY029486.1 | - | 1 - 2 | 40 | 77 | ||
| DA111448.1 | - | 1 - 2 | 75 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000741.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB030207.1,AK312139.1,AY029486.1,NM_016541.2 transcripts.
Set Membership: |
|

GNG13
Genomic Map