Assay Details
| Gene Symbol: ARHGEF38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242729.1 | NP_001229658.1 | 1 - 2 | 341 | 81 | |
| NM_017700.1 | NP_060170.1 | 1 - 2 | 341 | 81 | ||
| XM_011532051.2 | XP_011530353.1 | 1 - 2 | 356 | 81 | ||
| GenBank mRNA | AK000191.1 | - | 1 - 2 | 341 | 81 | |
| BC105695.1 | - | 1 - 2 | 329 | 40023595 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000544.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04007899.hg.1 | TC04000544.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007899.hg.1 | ENST00000265154.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007899.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007899.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007899.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007900.hg.1 | TC04000544.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007900.hg.1 | ENST00000265154.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007900.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007900.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007900.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007901.hg.1 | TC04000544.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007901.hg.1 | ENST00000265154.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007901.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007901.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007901.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188712.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188712.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188712.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188712.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188712.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188712.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188712.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188712.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188712.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188712.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188712.hg.1 | TC0400012829.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400188710.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188710.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188710.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188710.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188710.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188710.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188710.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188710.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188710.hg.1 | TC0400012829.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400188711.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188711.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188711.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188711.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188711.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188711.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188711.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188711.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188711.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188711.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188711.hg.1 | TC0400012829.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188709.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188709.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188709.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188709.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188709.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188709.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188709.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188709.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188709.hg.1 | TC0400012829.hg.1 | Clariom D TC |

ARHGEF38
Genomic Map