Assay Details
Gene Symbol: RETSAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017750.3 | NP_060220.3 | 5 - 6 | 1192 | 81 | |
GenBank mRNA | AK000303.1 | - | 3 - 4 | 421 | 81 | |
AK075261.1 | - | 5 - 6 | 1026 | 81 | ||
AK225305.1 | - | 3 - 4 | 421 | 81 | ||
AK296522.1 | - | 4 - 5 | 846 | 81 | ||
AL833237.1 | - | 5 - 6 | 1008 | 81 | ||
AY358568.1 | - | 5 - 6 | 1014 | 81 | ||
BC011418.1 | - | 3 - 4 | 522 | 81 | ||
BC068517.1 | - | 5 - 6 | 1022 | 4826703 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002037.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02032560.hg.1 | TC02002037.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02032560.hg.1 | ENST00000263854.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02032560.hg.1 | ENST00000295802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02032560.hg.1 | ENST00000457495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02032560.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02032556.hg.1 | TC02002037.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02032556.hg.1 | ENST00000263854.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02032556.hg.1 | ENST00000295802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02032556.hg.1 | ENST00000457495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02032556.hg.1 | ENST00000475624.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02032556.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200196838.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196838.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196838.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196838.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196838.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200196838.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196838.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196838.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196838.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196838.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200196838.hg.1 | TC0200013261.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200196839.hg.1 | TC0200013261.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200196837.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196837.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196837.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196837.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196837.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200196837.hg.1 | ENST00000295802.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196837.hg.1 | ENST00000429806.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196837.hg.1 | ENST00000438611.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196837.hg.1 | ENST00000449375.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200196837.hg.1 | NM_017750.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200196837.hg.1 | TC0200013261.hg.1 | Clariom D TC |
