Assay Details
| Gene Symbol: LINC00483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK000701.1 | - | 1 - 2 | 647 | 62 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000419688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000419688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000419688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000419688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | NR_073199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | NR_073199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | NR_073199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | NR_073199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | NR_073199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011117.hg.1 | NR_073199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000300458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000419688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000419688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000419688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000419688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000450727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000502517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | ENST00000576150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | NR_073199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | NR_073199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | NR_073199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | NR_073199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | NR_073199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011117.hg.1 | NR_073199.1 | Refseq |

LINC00483
Genomic Map