Assay Details
| Gene Symbol: AGPAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018361.3 | NP_060831.2 | 5 - 6 | 897 | 68 | |
| GenBank mRNA | AF375789.1 | - | 5 - 6 | 859 | 68 | |
| AK002072.1 | - | 5 - 6 | 561 | 68 | ||
| AL136587.1 | - | 5 - 6 | 587 | 68 | ||
| BC023550.2 | - | 5 - 6 | 841 | 4129973 | 68 | |
| BC068519.1 | - | 5 - 6 | 561 | 68 | ||
| BC080537.1 | - | 5 - 6 | 866 | 5015128 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000022.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000354.hg.1 | TC08000022.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000354.hg.1 | ENST00000285518.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000354.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000354.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | ENST00000533648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | ENST00000533648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144776.hg.1 | TC0800006566.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 114458 | - | - |

AGPAT5
Genomic Map