Assay Details
| Gene Symbol: BBX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020235.6 | NP_064620.2 | 13 - 14 | 2529 | 126 | |
| XM_011513003.2 | XP_011511305.1 | 13 - 14 | 2529 | 126 | ||
| XM_011513004.2 | XP_011511306.1 | 13 - 14 | 2657 | 126 | ||
| XM_011513005.2 | XP_011511307.1 | 13 - 14 | 2529 | 126 | ||
| XM_011513010.2 | XP_011511312.1 | 12 - 13 | 1520 | 126 | ||
| XM_017006882.1 | XP_016862371.1 | 14 - 15 | 2620 | 126 | ||
| XM_017006883.1 | XP_016862372.1 | 12 - 13 | 1520 | 126 | ||
| GenBank mRNA | AF168718.1 | - | 2 - 3 | 197 | 126 | |
| AF454941.1 | - | 13 - 14 | 2522 | 126 | ||
| AF454942.1 | - | 11 - 12 | 2402 | 126 | ||
| AK025944.1 | - | 3 - 4 | 780 | 126 | ||
| AL136769.1 | - | 11 - 12 | 2402 | 126 | ||
| AL834307.1 | - | 3 - 4 | 839 | 126 | ||
More Information
Set Membership: |
|

BBX
Genomic Map