| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006184.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006184.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006184.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006184.hg.1 |
TC20000457.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006185.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006185.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006185.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006185.hg.1 |
TC20000457.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006186.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006186.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006186.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006186.hg.1 |
TC20000457.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006183.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006183.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006183.hg.1 |
TC20000457.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006187.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006187.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006187.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006187.hg.1 |
TC20000457.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000154752.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154752.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154752.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154752.hg.1 |
TC2000007895.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000154752.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154752.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154752.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154755.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154755.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154755.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154755.hg.1 |
TC2000007895.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000154755.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154755.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154755.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154754.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154754.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154754.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154754.hg.1 |
TC2000007895.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000154754.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154754.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154754.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154750.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154750.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154750.hg.1 |
TC2000007895.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000154750.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154750.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154751.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154751.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154751.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154751.hg.1 |
TC2000007895.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000154751.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154751.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154751.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154753.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154753.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154753.hg.1 |
NM_020673.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154753.hg.1 |
TC2000007895.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000154753.hg.1 |
ENST00000244040.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154753.hg.1 |
ENST00000488949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154753.hg.1 |
NM_020673.2 |
Refseq |