| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000278780.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000278780.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000278780.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000278780.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000278780.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000278780.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000278780.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000278780.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000462208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000462208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000483661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000483661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000483661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000483661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000483661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000483661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000494030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000494030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000494030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000494030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
ENST00000494030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303462.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303462.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303462.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303462.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303462.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_001303462.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_021220.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_021220.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_021220.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_021220.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_021220.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_021220.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_021220.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009987.hg.1 |
NM_021220.3 |
Refseq |