Assay Details
| Gene Symbol: CHST8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022467.3 | NP_071912.2 | 1 - 2 | 672 | 71 | |
| GenBank mRNA | AF300612.1 | - | 1 - 2 | 133 | 71 | |
| AF305781.1 | - | 1 - 2 | 229 | 71 | ||
| BC018723.1 | - | 1 - 2 | 672 | 4137487 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000438.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF300612.1,AF305781.1,BC018723.1,NM_022467.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005690.hg.1 | ENST00000262622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005690.hg.1 | ENST00000434302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005690.hg.1 | ENST00000438847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005690.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005690.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005690.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005690.hg.1 | TC19000438.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005689.hg.1 | ENST00000262622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005689.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005689.hg.1 | TC19000438.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007783.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | TC1900007783.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157172.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | TC1900007783.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | ENST00000434302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | ENST00000438847.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | ENST00000591231.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | ENST00000604556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | NM_001127895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | NM_001127896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157173.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157171.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157171.hg.1 | NM_022467.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157171.hg.1 | TC1900007783.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900157171.hg.1 | ENST00000262622.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157171.hg.1 | NM_022467.3 | Refseq |

CHST8
Genomic Map