Assay Details
| Gene Symbol: ZNF649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023074.3 | NP_075562.2 | 1 - 2 | 140 | 110 | |
| GenBank mRNA | AK000909.1 | - | 1 - 2 | 76 | 110 | |
| AK022706.1 | - | 1 - 2 | 138 | 110 | ||
| AK225480.1 | - | 1 - 2 | 99 | 110 | ||
| AK315487.1 | - | 1 - 2 | 102 | 110 | ||
| AL713677.1 | - | 1 - 2 | 82 | 110 | ||
| BC005368.1 | - | 1 - 2 | 99 | 3930849 | 110 | |
| DA782250.1 | - | 1 - 2 | 140 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001789.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK022706.1,AK315487.1,AL713677.1,BC005368.1,NM_023074.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19024330.hg.1 | ENST00000354957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024330.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024330.hg.1 | TC19001789.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024331.hg.1 | ENST00000354957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024331.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024331.hg.1 | TC19001789.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000599671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000599671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000599671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000599671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024332.hg.1 | ENST00000354957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024332.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024332.hg.1 | TC19001789.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024329.hg.1 | ENST00000354957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024329.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024329.hg.1 | TC19001789.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000599671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | TC1900012019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000599671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207612.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | TC1900012019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207617.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | TC1900012019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207619.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000354957.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000595418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000596690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000597882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000599530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000599671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000599671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000599671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000599671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | ENST00000600738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012019.hg.1 | NM_023074.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 115165 | - | - |

ZNF649
Genomic Map