| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000374298.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000374298.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000374298.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000374298.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000374298.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000374298.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000374298.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000374298.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000481602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000481602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000538789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000538789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000538789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000538789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000538789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000538789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000538789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000606617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
ENST00000606617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_001287490.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_001287490.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_001287490.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_001287490.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_001287490.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_001287490.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_001287490.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_024037.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_024037.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_024037.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_024037.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_024037.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_024037.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_024037.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_024037.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013367.hg.1 |
NM_024037.2 |
Refseq |