Assay Details
| Gene Symbol: DCAF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005251577.3 | XP_005251634.1 | 4 - 5 | 1158 | 80 | |
| XM_017015128.1 | XP_016870617.1 | 3 - 4 | 960 | 80 | ||
| GenBank mRNA | BC003520.1 | - | 1 - 2 | 166 | 80 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

DCAF10
Genomic Map