Assay Details
| Gene Symbol: ASB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024701.3 | NP_078977.2 | 1 - 2 | 88 | 118 | |
| NR_024581.1 | 1 - 2 | 88 | 118 | |||
| NR_037164.1 | 1 - 2 | 88 | 118 | |||
| GenBank mRNA | AK023196.1 | - | 1 - 2 | 71 | 118 | |
| AK091935.1 | - | 1 - 2 | 88 | 118 | ||
| AK292071.1 | - | 1 - 2 | 74 | 118 | ||
| BC012056.1 | - | 1 - 2 | 65 | 4552643 | 118 | |
| BG436652.1 | - | 1 - 2 | 65 | 118 | ||
| BP249999.1 | - | 1 - 2 | 88 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001000.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK023196.1,AK292071.1,BC012056.1,NM_024701.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10012462.hg.1 | ENST00000357700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012462.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012462.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012462.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012462.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012462.hg.1 | TC10001000.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012463.hg.1 | ENST00000357700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012463.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012463.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012463.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012463.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012463.hg.1 | TC10001000.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166094.hg.1 | TC1000009677.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | ENST00000357700.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | ENST00000459912.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | ENST00000479033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | NM_024701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | NR_024581.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | NR_037164.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166095.hg.1 | TC1000009677.hg.1 | Clariom D TC |

ASB13
Genomic Map