Assay Details
| Gene Symbol: ADGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025048.3 | NP_079324.2 | 3 - 4 | 350 | 141 | |
| NM_153840.3 | NP_722582.2 | 3 - 4 | 350 | 141 | ||
| GenBank mRNA | AB045123.1 | - | 3 - 4 | 304 | 141 | |
| AK026337.1 | - | 3 - 4 | 281 | 141 | ||
| AK225460.1 | - | 3 - 4 | 281 | 141 | ||
| AL698860.1 | - | 3 - 4 | 334 | 141 | ||
| BC029358.1 | - | 2 - 3 | 175 | 141 | ||
| BC113634.1 | - | 2 - 3 | 163 | 141 | ||
| BC113636.1 | - | 2 - 3 | 163 | 141 | ||
| CD685935.1 | - | 3 - 4 | 344 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|

ADGRF1
Genomic Map