Assay Details
| Gene Symbol: LMAN2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142292.1 | NP_001135764.1 | 2 - 3 | 344 | 64 | |
| NM_001322346.1 | NP_001309275.1 | 2 - 3 | 360 | 64 | ||
| NM_001322354.1 | NP_001309283.1 | 2 - 3 | 374 | 64 | ||
| NM_001322356.1 | NP_001309285.1 | 2 - 3 | 360 | 64 | ||
| NM_030805.3 | NP_110432.1 | 2 - 3 | 344 | 64 | ||
| GenBank mRNA | AJ549957.1 | - | 2 - 3 | 308 | 64 | |
| AK075347.1 | - | 2 - 3 | 332 | 64 | ||
| AK222828.1 | - | 2 - 3 | 322 | 64 | ||
| AK299740.1 | - | 2 - 3 | 332 | 64 | ||
| AK308085.1 | - | 2 - 3 | 331 | 64 | ||
| AK310100.1 | - | 2 - 3 | 361 | 64 | ||
| AK312249.1 | - | 2 - 3 | 337 | 64 | ||
| AK316331.1 | - | 2 - 3 | 332 | 64 | ||
| AL136617.1 | - | 2 - 3 | 336 | 64 | ||
| AY358929.1 | - | 2 - 3 | 315 | 64 | ||
| BC000347.2 | - | 2 - 3 | 307 | 64 | ||
| BC005822.2 | - | 2 - 3 | 307 | 64 | ||
| BC005862.2 | - | 2 - 3 | 307 | 64 | ||
| BC067265.1 | - | 2 - 3 | 314 | 6502212 | 64 | |
| BX648002.1 | - | 2 - 3 | 322 | 64 | ||
| DA521101.1 | - | 2 - 3 | 321 | 64 | ||
| DC354352.1 | - | 2 - 3 | 346 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002111.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK299740.1,AK316331.1,NM_001322346.1,NM_001322354.1,NM_001322356.1 transcripts.
Set Membership: |
|

LMAN2L
Genomic Map