Assay Details
| Gene Symbol: SIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005068.2 | NP_005059.2 | 1 - 2 | 386 | 82 | |
| XM_005267100.2 | XP_005267157.1 | 2 - 3 | 962 | 82 | ||
| XM_011536072.2 | XP_011534374.1 | 2 - 3 | 962 | 82 | ||
| XM_017011197.1 | XP_016866686.1 | 2 - 3 | 694 | 82 | ||
| GenBank mRNA | U70212.1 | - | 1 - 2 | 395 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001969.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ureteric bud development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
nervous system development
cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein heterodimerization activity
ureteric bud development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
nervous system development
cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06024380.hg.1 | TC06001969.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024380.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024380.hg.1 | ENST00000369208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024380.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000505753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000505753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024378.hg.1 | TC06001969.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024378.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024378.hg.1 | ENST00000369208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024378.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187387.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187387.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187387.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187387.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187387.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187387.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187387.hg.1 | TC0600012670.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600187389.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187389.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187389.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187389.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187389.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187389.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187389.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187389.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187389.hg.1 | TC0600012670.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600187390.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187390.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187390.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187390.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187390.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187390.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187390.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187390.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187390.hg.1 | TC0600012670.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000262901.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000369208.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000505753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000505753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | ENST00000511871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012670.hg.1 | NM_005068.2 | Refseq |

SIM1
Genomic Map