| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011201.hg.1 |
TC06000960.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011201.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011201.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011202.hg.1 |
TC06000960.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011202.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011202.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011203.hg.1 |
TC06000960.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011203.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011203.hg.1 |
ENST00000368365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011203.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164222.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164222.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164222.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164222.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164222.hg.1 |
TC0600009362.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600164225.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164225.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164225.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164225.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164225.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164225.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164225.hg.1 |
TC0600009362.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
ENST00000368365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009362.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164223.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164223.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164223.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164223.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164223.hg.1 |
TC0600009362.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600164224.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164224.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164224.hg.1 |
ENST00000368364.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164224.hg.1 |
NM_012259.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164224.hg.1 |
TC0600009362.hg.1 |
Clariom D TC |