Assay Details
| Gene Symbol: IL12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002187.2 | NP_002178.2 | 4 - 5 | 524 | 129 | |
| GenBank mRNA | AF180563.1 | - | 3 - 4 | 488 | 129 | |
| BC067498.1 | - | 3 - 4 | 486 | 6971868 | 129 | |
| BC067499.1 | - | 3 - 4 | 486 | 6971869 | 129 | |
| BC067500.1 | - | 3 - 4 | 486 | 6971872 | 129 | |
| BC067501.1 | - | 3 - 4 | 486 | 6971873 | 129 | |
| BC067502.1 | - | 3 - 4 | 486 | 6971874 | 129 | |
| BC074723.2 | - | 4 - 5 | 513 | 30915173 | 129 | |
| M65272.1 | - | 4 - 5 | 524 | 129 | ||
| M65290.1 | - | 3 - 4 | 495 | 129 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001997.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (17)
Expression of the voltage-gated sodium channel NaV1.5 in the macrophage late endosome regulates endosomal acidification.
- Authors
- Carrithers MD, Dib-Hajj S, Carrithers LM, Tokmoulina G, Pypaert M, Jonas EA, Waxman SG
- Journal
- J Immunol - Page No 7822 (Fri Jun 15 00:00:00 EDT 2007)
- Volume (Year)
- 178 (2007)
- Product Usage
- -
Adjuvant activity on murine and human macrophages.
- Authors
- Quesniaux V, Erard F, Ryffel B
- Journal
- Methods Mol Biol - Page No 117 (Fri Jan 01 00:00:00 EST 2010)
- Volume (Year)
- 626 (2010)
- Product Usage
- -
Multiple signaling pathways contribute to synergistic TLR ligand-dependent cytokine gene expression in human monocyte-derived macrophages and dendritic cells.
- Authors
- Makela SM, Strengell M, Pietila TE, Osterlund P, Julkunen I
- Journal
- J Leukoc Biol - Page No 664 (Wed Apr 01 00:00:00 EDT 2009)
- Volume (Year)
- 85 (2009)
- Product Usage
- -
Inflammatory and microRNA Gene Expression as Prognostic Classifiers of Barrett's Associated Esophageal Adenocarcinoma.
- Authors
- Nguyen GH, Schetter AJ, Chou DB, Bowman ED, Zhao R, Hawkes JE, Mathe E, Kumamoto K, Zhao Y, Budhu A, Hagiwara N, Wang XW, Miyashita M, Casson AG, Harris CC
- Journal
- Clin Cancer Res - Page No (Mon Oct 25 00:00:00 EDT 2010)
- Volume (Year)
- (2010)
- Product Usage
- -
Differences in the inducible gene expression and protein production of IL-12p40, IL-12p70 and IL-23: involvement of p38 and JNK kinase pathways.
- Authors
- Dobreva ZG, Stanilova SA, Miteva LD
- Journal
- Cytokine - Page No 76 (Tue Jul 01 00:00:00 EDT 2008)
- Volume (Year)
- 43 (2008)
- Product Usage
- -
Innate immune responses to TREM-1 activation: overlap, divergence, and positive and negative cross-talk with bacterial lipopolysaccharide.
- Authors
- Dower K, Ellis DK, Saraf K, Jelinsky SA, Lin LL
- Journal
- J Immunol - Page No 3520 (Sat Mar 01 00:00:00 EST 2008)
- Volume (Year)
- 180 (2008)
- Product Usage
- -
Expression of cytokine-associated genes in dendritic cells (DCs): comparison between adult peripheral blood- and umbilical cord blood-derived DCs by cDNA microarray.
- Authors
- Koga Y, Matsuzaki A, Suminoe A, Hattori H, Hara T
- Journal
- Immunol Lett - Page No 55 (Fri Feb 15 00:00:00 EST 2008)
- Volume (Year)
- 116 (2008)
- Product Usage
- -
Acquisition of hemozoin by monocytes down-regulates interleukin-12 p40 (IL-12p40) transcripts and circulating IL-12p70 through an IL-10-dependent mechanism: in vivo and in vitro findings in severe malarial anemia.
- Authors
- Keller CC, Yamo O, Ouma C, Ong'echa JM, Ounah D, Hittner JB, Vulule JM, Perkins DJ
- Journal
- Infect Immun - Page No 5249 (Fri Sep 01 00:00:00 EDT 2006)
- Volume (Year)
- 74 (2006)
- Product Usage
- -
Postmenopausal expression changes of immune system-related genes in human bone tissue.
- Authors
- Kosa JP, Balla B, Kiss J, Podani J, Takacs I, Lazary A, Nagy Z, Bacsi K, Karsai A, Speer G, Lakatos P
- Journal
- J Clin Immunol - Page No 761 (Sun Nov 01 00:00:00 EDT 2009)
- Volume (Year)
- 29 (2009)
- Product Usage
- -
Characterization of the innate immune signalling pathways in hepatocyte cell lines.
- Authors
- Preiss S, Thompson A, Chen X, Rodgers S, Markovska V, Desmond P, Visvanathan K, Li K, Locarnini S, Revill P
- Journal
- J Viral Hepat - Page No 888 (Mon Dec 01 00:00:00 EST 2008)
- Volume (Year)
- 15 (2008)
- Product Usage
- -
Interactions among type I and type II interferon, tumor necrosis factor, and beta-estradiol in the regulation of immune response-related gene expressions in systemic lupus erythematosus.
- Authors
- Lee HM, Mima T, Sugino H, Aoki C, Adachi Y, Yoshio-Hoshino N, Matsubara K, Nishimoto N
- Journal
- Arthritis Res Ther - Page No R1 (Thu Jan 01 00:00:00 EST 2009)
- Volume (Year)
- 11 (2009)
- Product Usage
- -
Helicobacter pylori stimulates dendritic cells to induce interleukin-17 expression from CD4+ T lymphocytes.
- Authors
- Khamri W, Walker MM, Clark P, Atherton JC, Thursz MR, Bamford KB, Lechler RI, Lombardi G
- Journal
- Infect Immun - Page No 845 (Mon Feb 01 00:00:00 EST 2010)
- Volume (Year)
- 78 (2010)
- Product Usage
- -
Using distinct molecular signatures of human monocytes and dendritic cells to predict adjuvant activity and pyrogenicity of TLR agonists.
- Authors
- Kamgang RK, Ramos I, Rodrigues Duarte L, Ghielmetti M, Freudenberg M, Dahinden C, Padovan E
- Journal
- Med Microbiol Immunol - Page No 369 (Mon Dec 01 00:00:00 EST 2008)
- Volume (Year)
- 197 (2008)
- Product Usage
- -
JNK1 and JNK2 differently regulate IL-12 production in THP-1 macrophage cells.
- Authors
- Utsugi M, Dobashi K, Ono A, Ishizuka T, Hisada T, Koga Y, Shimizu Y, Kawata T, Matsuzaki S, Aoki H, Kamide Y, Mori M
- Journal
- Cytokine - Page No 127 (Sun Aug 01 00:00:00 EDT 2010)
- Volume (Year)
- 51 (2010)
- Product Usage
- -
High expression of Foxp3, IL-23p19 and survivin mRNA in colorectal carcinoma.
- Authors
- Stanilov N, Miteva L, Mintchev N, Stanilova S
- Journal
- Int J Colorectal Dis - Page No 151 (Sun Feb 01 00:00:00 EST 2009)
- Volume (Year)
- 24 (2009)
- Product Usage
- -
An analysis of select pathogenic messages in lesional and non-lesional psoriatic skin using non-invasive tape harvesting.
- Authors
- Benson NR, Papenfuss J, Wong R, Motaal A, Tran V, Panko J, Krueger GG
- Journal
- J Invest Dermatol - Page No 2234 (Sun Oct 01 00:00:00 EDT 2006)
- Volume (Year)
- 126 (2006)
- Product Usage
- -
Differential expression of immunoregulatory genes in monocytes in response to Porphyromonas gingivalis and Escherichia coli lipopolysaccharide.
- Authors
- Barksby HE, Nile CJ, Jaedicke KM, Taylor JJ, Preshaw PM
- Journal
- Clin Exp Immunol - Page No 479 (Mon Jun 01 00:00:00 EDT 2009)
- Volume (Year)
- 156 (2009)
- Product Usage
- -

IL12B
Genomic Map