| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16005571.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16005571.hg.1 |
ENST00000444228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16005571.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16005571.hg.1 |
TC16000376.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600155274.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155274.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155274.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600155274.hg.1 |
ENST00000389202.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155274.hg.1 |
ENST00000444228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155274.hg.1 |
NM_005353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600155274.hg.1 |
TC1600007532.hg.1 |
Clariom D TC |