Assay Details
| Gene Symbol: DAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199849.1 | NP_001186778.1 | 4 - 5 | 351 | 60 | |
| NM_004632.3 | NP_004623.1 | 3 - 4 | 255 | 60 | ||
| NM_033657.2 | NP_387506.1 | 3 - 4 | 324 | 60 | ||
| XM_011509972.2 | XP_011508274.1 | 3 - 4 | 254 | 60 | ||
| XM_017002289.1 | XP_016857778.1 | 3 - 4 | 323 | 60 | ||
| XM_017002290.1 | XP_016857779.1 | 4 - 5 | 347 | 60 | ||
| GenBank mRNA | AK225869.1 | - | 4 - 5 | 351 | 60 | |
| AK293117.1 | - | 3 - 4 | 256 | 60 | ||
| BC002358.1 | - | 2 - 3 | 283 | 60 | ||
| BC107487.1 | - | 3 - 4 | 255 | 40032516 | 60 | |
| BC107488.1 | - | 3 - 4 | 324 | 40032512 | 60 | |
| BT019494.1 | - | 2 - 3 | 167 | 60 | ||
| CR749790.1 | - | 3 - 4 | 335 | 60 | ||
| U18321.1 | - | 3 - 4 | 336 | 60 | ||
| X83544.1 | - | 3 - 4 | 240 | 60 | ||
More Information
Set Membership: |
|

DAP3
Genomic Map