| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000311852.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000547074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000547074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000547074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000547074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000547279.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000547279.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000547279.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000547596.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000547596.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000547596.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
ENST00000548761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006659.hg.1 |
NM_004995.3 |
Refseq |