Assay Details
| Gene Symbol: TACSTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002353.2 | NP_002344.2 | 1 | 80 | 64 | |
| GenBank mRNA | AW052118.1 | - | 1 | 64 | ||
| X77753.1 | - | 1 | 357 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002706.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_002353.2,X77753.1 transcripts.
Set Membership: |
|

TACSTD2
Genomic Map