| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
TC04001820.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
ENST00000284776.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
ENST00000319471.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
ENST00000355634.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
ENST00000393528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
ENST00000431808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
ENST00000437304.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
ENST00000448662.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
ENST00000449407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
NM_001145670.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
NM_001145671.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
NM_001145672.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
NM_001145673.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
NM_003603.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04024984.hg.1 |
NM_021069.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000284776.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000319454.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000319471.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000355634.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000393523.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000393528.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000414724.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000415274.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000419063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000420158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000421420.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000421639.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000425679.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000428330.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000429056.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000430503.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000431902.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000432655.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000435480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000437304.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000439049.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000439914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000444771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000444781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000445115.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000445343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000448662.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000449407.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000450341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000451701.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000451958.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000452351.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000456596.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000457247.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000457934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000463104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000464819.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000464975.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000470685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000478249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000493709.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_001145670.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_001145671.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_001145672.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_001145673.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_001270771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_003603.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_021069.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000284776.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000319454.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000319471.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000355634.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000393523.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000393528.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000414724.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000415274.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000419063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000420158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000421420.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000421639.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000425679.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000428330.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000429056.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000430503.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000431902.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000432655.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000435480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000437304.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000439049.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000439914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000444771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000444781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000445115.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000445343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000448662.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000449407.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000450341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000451701.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000451958.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000452351.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000456596.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000457247.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000457934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000463104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000464819.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000464975.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000470685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000478249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
ENST00000493709.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_001145670.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_001145671.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_001145672.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_001145673.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_001270771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_003603.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
NM_021069.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186749.hg.1 |
TC0400012644.hg.1 |
Clariom D TC |