Assay Details
| Gene Symbol: DCTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004082.4 | NP_004073.2 | 5 - 6 | 733 | 88 | |
| GenBank mRNA | BX640799.1 | - | 5 - 6 | 512 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCTN1| Gene Symbol: DCTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004082.4 | NP_004073.2 | 5 - 6 | 733 | 88 | |
| GenBank mRNA | BX640799.1 | - | 5 - 6 | 512 | 88 | |
Set Membership: |
|