Assay Details
| Gene Symbol: UTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006786.3 | NP_006777.1 | 1 - 2 | 146 | 102 | |
| XM_011540538.1 | XP_011538840.1 | 2 - 3 | 304 | 102 | ||
| XM_017000117.1 | XP_016855606.1 | 2 - 3 | 304 | 102 | ||
| GenBank mRNA | AF104118.1 | - | 1 - 2 | 137 | 102 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002162.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
negative regulation of glomerular filtration
muscle contraction
positive regulation of cytosolic calcium ion concentration
chemical synaptic transmission
regulation of blood pressure
negative regulation of heart rate
positive regulation of heart rate
positive regulation of fibroblast migration
positive regulation of circadian sleep/wake cycle, wakefulness
positive regulation of synaptic transmission, cholinergic
positive regulation of collagen biosynthetic process
response to testosterone
negative regulation of urine volume
negative regulation of renal sodium excretion
response to drug
positive regulation of cell differentiation
positive regulation of angiogenesis
negative regulation of blood pressure
positive regulation of blood pressure
positive regulation of vasodilation
positive regulation of circadian sleep/wake cycle, REM sleep
negative regulation of insulin secretion
positive regulation of fibroblast proliferation
receptor binding
hormone activity
response to hypoxia
negative regulation of glomerular filtration
muscle contraction
positive regulation of cytosolic calcium ion concentration
chemical synaptic transmission
regulation of blood pressure
negative regulation of heart rate
positive regulation of heart rate
positive regulation of fibroblast migration
positive regulation of circadian sleep/wake cycle, wakefulness
positive regulation of synaptic transmission, cholinergic
positive regulation of collagen biosynthetic process
response to testosterone
negative regulation of urine volume
negative regulation of renal sodium excretion
response to drug
positive regulation of cell differentiation
positive regulation of angiogenesis
negative regulation of blood pressure
positive regulation of blood pressure
positive regulation of vasodilation
positive regulation of circadian sleep/wake cycle, REM sleep
negative regulation of insulin secretion
positive regulation of fibroblast proliferation
receptor binding
hormone activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01033370.hg.1 | TC01002162.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033370.hg.1 | ENST00000361696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033370.hg.1 | ENST00000377516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033370.hg.1 | ENST00000400910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033370.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033368.hg.1 | TC01002162.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033368.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033368.hg.1 | ENST00000361696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033368.hg.1 | ENST00000377516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033368.hg.1 | ENST00000400910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033368.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033368.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194256.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194256.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194256.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194256.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194256.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194256.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194256.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194256.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194256.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194256.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194256.hg.1 | TC0100012732.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100194258.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194258.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194258.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194258.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194258.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194258.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194258.hg.1 | TC0100012732.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000054668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012732.hg.1 | NM_021995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194259.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194259.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194259.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194259.hg.1 | ENST00000361696.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194259.hg.1 | ENST00000377516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194259.hg.1 | NM_006786.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194259.hg.1 | TC0100012732.hg.1 | Clariom D TC |

UTS2
Genomic Map