Assay Details
| Gene Symbol: BNIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205.2 | NP_001196.2 | 3 - 4 | 376 | 70 | |
| NM_013979.2 | NP_053582.2 | 4 - 5 | 505 | 70 | ||
| XM_011534638.1 | XP_011532940.1 | 4 - 5 | 500 | 70 | ||
| XM_011534639.1 | XP_011532941.1 | 4 - 5 | 500 | 70 | ||
| GenBank mRNA | AF083957.1 | - | 4 - 5 | 411 | 70 | |
| AU311051.1 | - | 4 - 5 | 505 | 70 | ||
| BC010959.1 | - | 3 - 4 | 301 | 4052817 | 70 | |
| U15172.1 | - | 3 - 4 | 282 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000959.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05013958.hg.1 | TC05000959.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013958.hg.1 | ENST00000231668.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013958.hg.1 | ENST00000351486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013958.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013958.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000519459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000519459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000519459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000519459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166422.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166422.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166422.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166422.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166422.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166422.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166422.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166422.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166422.hg.1 | TC0500009490.hg.1 | Clariom D TC |

BNIP1
Genomic Map