Assay Details
| Gene Symbol: MBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018328.4 | NP_060798.2 | 9 | 2906 | 93 | |
| XM_011511470.2 | XP_011509772.1 | 9 | 3138 | 93 | ||
| XM_011511472.2 | XP_011509774.1 | 7 | 2718 | 93 | ||
| XM_017004482.1 | XP_016859971.1 | 5 | 2588 | 93 | ||
| XM_017004483.1 | XP_016859972.1 | 5 | 2588 | 93 | ||
| XM_017004484.1 | XP_016859973.1 | 5 | 2588 | 93 | ||
| XM_017004485.1 | XP_016859974.1 | 5 | 2588 | 93 | ||
| GenBank mRNA | AB040894.1 | - | 4 | 2199 | 93 | |
| AK304424.1 | - | 4 | 2221 | 93 | ||
| BC014534.1 | - | 1 | 355 | 93 | ||
| BC150264.1 | - | 4 | 2199 | 9007212 | 93 | |
| EF542797.1 | - | 9 | 2897 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

MBD5
Genomic Map