Assay Details
| Gene Symbol: SLC25A37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF116640.1 | - | 1 | 369 | 118 | |
| AK093931.1 | - | 1 | 953 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF116640.1 transcripts.
Set Membership: |
|

SLC25A37
Genomic Map