| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
ENST00000593537.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
ENST00000593537.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
ENST00000593537.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
ENST00000593537.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
ENST00000594938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
ENST00000594938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
ENST00000595857.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
ENST00000595857.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
ENST00000595857.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
ENST00000595857.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
NM_006179.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
NM_006179.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
NM_006179.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011998.hg.1 |
NM_006179.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
ENST00000593537.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
ENST00000593537.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
ENST00000593537.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
ENST00000593537.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
ENST00000594938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
ENST00000594938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
ENST00000595857.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
ENST00000595857.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
ENST00000595857.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
ENST00000595857.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
NM_006179.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
NM_006179.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
NM_006179.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011998.hg.1 |
NM_006179.4 |
Refseq |