Assay Details
| Gene Symbol: IFNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024013.2 | NP_076918.1 | 1 | 637 | 115 | |
| GenBank mRNA | AB578886.1 | - | 1 | 115 | ||
| V00537.1 | - | 1 | 639 | 115 | ||
| V00538.1 | - | 1 | 115 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000103.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Role of the long cytoplasmic domain of the SIV Env glycoprotein in early and late stages of infection.
- Authors
- Vzorov AN, Weidmann A, Kozyr NL, Khaoustov V, Yoffe B, Compans RW
- Journal
- Retrovirology - Page No 94 (Mon Jan 01 00:00:00 EST 2007)
- Volume (Year)
- 4 (2007)
- Product Usage
- -

IFNA1
Genomic Map