Assay Details
| Gene Symbol: CAAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167575.1 | NP_001161047.1 | 4 - 5 | 934 | 80 | |
| NM_024828.3 | NP_079104.3 | 4 - 5 | 780 | 80 | ||
| GenBank mRNA | AK023719.1 | - | 4 - 5 | 754 | 80 | |
| AK300739.1 | - | 4 - 5 | 548 | 80 | ||
| AK301673.1 | - | 4 - 5 | 478 | 80 | ||
| BC014658.1 | - | 4 - 5 | 756 | 4648932 | 80 | |
| BC071953.1 | - | 4 - 5 | 679 | 5431506 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000982.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000444762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000517946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000517946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000517946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000517946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012886.hg.1 | TC09000982.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012886.hg.1 | ENST00000333916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012886.hg.1 | ENST00000495958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012886.hg.1 | ENST00000535437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012886.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012886.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900166777.hg.1 | TC0900009768.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000333916.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000444762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000483493.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000495958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000517946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000517946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000517946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000517946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000520187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | ENST00000625311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_001167575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009768.hg.1 | NM_024828.3 | Refseq |

CAAP1
Genomic Map