Assay Details
| Gene Symbol: PCDH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032456.2 | NP_115832.1 | 1 | 4334 | 115 | |
| GenBank mRNA | AB006756.1 | - | 1 | 3886 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

PCDH7
Genomic Map