Assay Details
| Gene Symbol: RNASE7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ131212.1 | - | 1 - 2 | 623 | 60 | |
| AJ306608.1 | - | 2 - 3 | 789 | 60 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000298690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | ENST00000481538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | NM_032572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006548.hg.1 | NM_032572.3 | Refseq |

RNASE7
Genomic Map