Assay Details
| Gene Symbol: USMG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032747.3 | NP_116136.1 | 1 - 2 | 108 | 63 | |
| GenBank mRNA | BC007087.1 | - | 1 - 2 | 61 | 4245636 | 63 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001630.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC007087.1,NM_032747.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10021295.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021295.hg.1 | ENST00000369825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021295.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021295.hg.1 | TC10001630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021301.hg.1 | ENST00000309579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021301.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021301.hg.1 | ENST00000369815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021301.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021301.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021301.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021301.hg.1 | TC10001630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000309579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000309579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000309579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000309579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | ENST00000408840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011716.hg.1 | NR_031707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021305.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021305.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021305.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021305.hg.1 | TC10001630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021296.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021296.hg.1 | ENST00000369825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021296.hg.1 | ENST00000408840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021296.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021296.hg.1 | NR_031707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021296.hg.1 | TC10001630.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | ENST00000309579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | ENST00000309579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180205.hg.1 | TC1000011716.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000180209.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180209.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180209.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180209.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180209.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180209.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180209.hg.1 | TC1000011716.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000309579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000309579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000309579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000309579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | ENST00000408840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_001206426.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_001206427.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011716.hg.1 | NR_031707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180199.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180199.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180199.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180199.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180199.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180199.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180199.hg.1 | TC1000011716.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000180200.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180200.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180200.hg.1 | ENST00000408840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180200.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180200.hg.1 | NR_031707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180200.hg.1 | ENST00000337003.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180200.hg.1 | ENST00000369825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180200.hg.1 | ENST00000408840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180200.hg.1 | NM_032747.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180200.hg.1 | NR_031707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180200.hg.1 | TC1000011716.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 200000 | - | - |

USMG5
Genomic Map