Assay Details
| Gene Symbol: GAL3ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033036.2 | NP_149025.1 | 1 - 2 | 173 | 100 | |
| GenBank mRNA | AB053232.1 | - | 1 - 2 | 173 | 100 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001953.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB053232.1,NM_033036.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
monosaccharide metabolic process
sulfur compound metabolic process
glycolipid biosynthetic process
oligosaccharide metabolic process
proteoglycan biosynthetic process
poly-N-acetyllactosamine metabolic process
galactosylceramide sulfotransferase activity
carbohydrate binding
3'-phosphoadenosine 5'-phosphosulfate binding
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
monosaccharide metabolic process
sulfur compound metabolic process
glycolipid biosynthetic process
oligosaccharide metabolic process
proteoglycan biosynthetic process
poly-N-acetyllactosamine metabolic process
galactosylceramide sulfotransferase activity
carbohydrate binding
3'-phosphoadenosine 5'-phosphosulfate binding
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11023450.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023450.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023450.hg.1 | TC11001953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023447.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023447.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023447.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023447.hg.1 | TC11001953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023448.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023448.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023448.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023448.hg.1 | TC11001953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186687.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186687.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186687.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186687.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186687.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186687.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186687.hg.1 | TC1100011270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100186688.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186688.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186688.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186688.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186688.hg.1 | TC1100011270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100186684.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186684.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186684.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186684.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186684.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186684.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186684.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186684.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186684.hg.1 | TC1100011270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011270.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186685.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186685.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186685.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186685.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186685.hg.1 | ENST00000312006.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186685.hg.1 | ENST00000527048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186685.hg.1 | ENST00000527878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186685.hg.1 | NM_033036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186685.hg.1 | TC1100011270.hg.1 | Clariom D TC |

GAL3ST3
Genomic Map