Assay Details
| Gene Symbol: GFOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF264036.1 | - | 1 - 3 | 177 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF264036.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000605067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000605067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000605067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NR_038459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NR_038459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010853.hg.1 | NR_038459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379278.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379278.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379278.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000379287.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000603223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000605067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000605067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000605067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | ENST00000612338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_001242630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NM_018988.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NR_038459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NR_038459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010853.hg.1 | NR_038459.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 207236 | - | - |

GFOD1
Genomic Map