Assay Details
| Gene Symbol: TIGD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF251050.2 | - | 1 - 2 | 2301 | 75 | |
| Gene Symbol: ZNF263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011522344.1 | XP_011520646.1 | 6 - 7 | 75 | ||
| XM_017022850.1 | XP_016878339.1 | 5 - 6 | 75 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000572748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000572748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000572748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000572748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000574253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000574253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000574253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000574253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000574674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000574674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000574674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000575332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000575332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000575332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000575332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000575332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000575823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000575823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000575823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | ENST00000575823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000219069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000572748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000572748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000572748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000572748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000573578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000574253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000574253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000574253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000574253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000574674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000574674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000574674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000575332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000575332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000575332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000575332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000575332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000575823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000575823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000575823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | ENST00000575823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011339.hg.1 | NM_005741.4 | Refseq |

TIGD7
Genomic Map