Assay Details
| Gene Symbol: PPP1R3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033215.4 | NP_149992.3 | 1 - 2 | 1038 | 78 | |
| XM_005272687.3 | XP_005272744.1 | 1 - 2 | 1059 | 78 | ||
| XM_017029934.1 | XP_016885423.1 | 1 - 2 | 1059 | 78 | ||
| XM_017029935.1 | XP_016885424.1 | 1 - 2 | 1059 | 78 | ||
| GenBank mRNA | AK056909.1 | - | 1 - 2 | 863 | 78 | |
| AK303738.1 | - | 2 - 3 | 364 | 78 | ||
| DC412626.1 | - | 2 - 3 | 366 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000270.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK303738.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003526.hg.1 | TC0X000270.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003526.hg.1 | ENST00000055335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003526.hg.1 | ENST00000438316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003526.hg.1 | ENST00000466508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003526.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003524.hg.1 | TC0X000270.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003524.hg.1 | ENST00000055335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003524.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003529.hg.1 | TC0X000270.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003529.hg.1 | ENST00000055335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003529.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003529.hg.1 | ENST00000438316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003529.hg.1 | ENST00000466508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003529.hg.1 | ENST00000495799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003529.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003529.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177433.hg.1 | TC0X00011286.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00177430.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177430.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177430.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177430.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177430.hg.1 | TC0X00011286.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00177431.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177431.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177431.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177431.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177431.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177431.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177431.hg.1 | TC0X00011286.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00177429.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177429.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177429.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177429.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177429.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177429.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177429.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177429.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177429.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177429.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177429.hg.1 | TC0X00011286.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000055335.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000376188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000466508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | ENST00000495799.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_001184745.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_033215.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011286.hg.1 | NM_033215.4 | Refseq |

PPP1R3F
Genomic Map