Assay Details
| Gene Symbol: TBL1Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033284.1 | NP_150600.1 | 2 - 3 | 378 | 65 | |
| XM_017030086.1 | XP_016885575.1 | 2 - 3 | 367 | 65 | ||
| XM_017030087.1 | XP_016885576.1 | 2 - 3 | 367 | 65 | ||
| GenBank mRNA | AF332220.1 | - | 2 - 3 | 378 | 65 | |
| AK298210.1 | - | 2 - 3 | 367 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF332220.1, AK298210.1, NM_033284.1, XM_017030086.1, XM_017030087.1 transcripts.
Set Membership: |
|

TBL1Y
Genomic Map