Assay Details
| Gene Symbol: ALDH1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK225950.1 | - | 2 | 258 | 85 | |
| AK313344.1 | - | 2 | 308 | 85 | ||
| BC001619.1 | - | 2 | 282 | 3356389 | 85 | |
| BE889810.1 | - | 315 | 85 | |||
| BT007418.1 | - | 1 | 192 | 85 | ||
| DB256700.1 | - | 2 | 359 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

ALDH1B1
Genomic Map