Assay Details
| Gene Symbol: ANG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097577.2 | NP_001091046.1 | 2 | 283 | 91 | |
| NM_001145.4 | NP_001136.1 | 2 | 763 | 91 | ||
| GenBank mRNA | AK313989.1 | - | 2 | 349 | 91 | |
| AV646929.1 | - | 2 | 91 | |||
| BC020704.1 | - | 2 | 276 | 4716144 | 91 | |
| BC054880.1 | - | 2 | 330 | 6453640 | 91 | |
| BC062698.1 | - | 2 | 282 | 4716820 | 91 | |
| BQ773604.1 | - | 1 | 91 | |||
| CB157060.1 | - | 1 | 91 | |||
| CB963791.1 | - | 2 | 354 | 91 | ||
| CR407633.1 | - | 1 | 163 | 91 | ||
| FJ236304.1 | - | 2 | 763 | 91 | ||
| Gene Symbol: RNASE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CB051790.1 | - | 1 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC020704.1 transcripts.
Set Membership: |
|

ANG
Genomic Map