Assay Details
| Gene Symbol: TNFRSF10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003844.3 | NP_003835.3 | 1 - 2 | 413 | 83 | |
| GenBank mRNA | AK291299.1 | - | 1 - 2 | 330 | 83 | |
| BC012866.1 | - | 1 - 2 | 372 | 3857315 | 83 | |
| BT006906.1 | - | 1 - 2 | 307 | 83 | ||
| DB101031.1 | - | 1 - 2 | 413 | 83 | ||
| FJ795027.1 | - | 1 - 2 | 306 | 83 | ||
| U90875.1 | - | 1 - 2 | 307 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001052.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
inflammatory response
immune response
signal transduction
cell surface receptor signaling pathway
activation of NF-kappaB-inducing kinase activity
multicellular organism development
extrinsic apoptotic signaling pathway via death domain receptors
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
TRAIL-activated apoptotic signaling pathway
regulation of cell proliferation
regulation of apoptotic process
cellular response to mechanical stimulus
extrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
protease binding
receptor activity
tumor necrosis factor-activated receptor activity
death receptor activity
protein binding
transcription factor binding
TRAIL binding
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
inflammatory response
immune response
signal transduction
cell surface receptor signaling pathway
activation of NF-kappaB-inducing kinase activity
multicellular organism development
extrinsic apoptotic signaling pathway via death domain receptors
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
TRAIL-activated apoptotic signaling pathway
regulation of cell proliferation
regulation of apoptotic process
cellular response to mechanical stimulus
extrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
protease binding
receptor activity
tumor necrosis factor-activated receptor activity
death receptor activity
protein binding
transcription factor binding
TRAIL binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000519862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000519862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000519862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013394.hg.1 | TC08001052.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013394.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013391.hg.1 | TC08001052.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013391.hg.1 | ENST00000221132.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013391.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013393.hg.1 | TC08001052.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013393.hg.1 | ENST00000221132.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013393.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000519862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000519862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000519862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009875.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167059.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167059.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167059.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167059.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167059.hg.1 | TC0800009875.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800167057.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167057.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167057.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167057.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167057.hg.1 | TC0800009875.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800167058.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167058.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167058.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167058.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167058.hg.1 | ENST00000613472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167058.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167058.hg.1 | TC0800009875.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800167060.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167060.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167060.hg.1 | TC0800009875.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800167055.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167055.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167055.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167055.hg.1 | ENST00000221132.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167055.hg.1 | ENST00000524158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167055.hg.1 | NM_003844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167055.hg.1 | TC0800009875.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 129544 | - | - |

TNFRSF10A
Genomic Map