Assay Details
| Gene Symbol: HAVCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173393.2 | NP_001166864.1 | 1 - 2 | 203 | 63 | |
| GenBank mRNA | AF043724.1 | - | 1 - 2 | 43 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001977.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF043724.1,NM_001173393.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010234.hg.1 | ENST00000598671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010234.hg.1 | NR_003603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010234.hg.1 | NR_003603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000339252.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000517644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000517644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000517644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000517644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000518745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000518745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000518745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000518745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000522693.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000523175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000523175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000523175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000523175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000523175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000523175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000523175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000523175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000523175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000523175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | ENST00000625904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001173393.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001173393.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001173393.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001173393.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001173393.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001173393.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001173393.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001173393.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001173393.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001173393.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001173393.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_001308156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012588.hg.1 | NM_012206.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010234.hg.1 | ENST00000598671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010234.hg.1 | NR_003603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010234.hg.1 | NR_003603.1 | Refseq |

HAVCR1
Genomic Map