Assay Details
| Gene Symbol: ROM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000327.3 | NP_000318.1 | 1 - 2 | 1136 | 85 | |
| GenBank mRNA | AK313674.1 | - | 1 - 2 | 1136 | 85 | |
| BC008100.1 | - | 1 - 2 | 726 | 4177529 | 85 | |
| BC036268.1 | - | 1 - 2 | 726 | 85 | ||
| L07894.1 | - | 1 - 2 | 600 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000557.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000525801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000525801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000525801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000525801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000529273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000529273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006743.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006743.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006743.hg.1 | TC11000557.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006744.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006744.hg.1 | ENST00000534093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006744.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006744.hg.1 | TC11000557.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006742.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006742.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006742.hg.1 | TC11000557.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100155966.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155966.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155966.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155966.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155966.hg.1 | TC1100007863.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100155970.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155970.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155970.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155970.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155970.hg.1 | TC1100007863.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100155967.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155967.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155967.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155967.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155967.hg.1 | TC1100007863.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000525801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000525801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000525801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000525801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000529273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000529273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007863.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155969.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155969.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155969.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155969.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155969.hg.1 | TC1100007863.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100155973.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155973.hg.1 | ENST00000525801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155973.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155973.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155973.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155973.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155973.hg.1 | ENST00000525801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155973.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155973.hg.1 | ENST00000534093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155973.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155973.hg.1 | TC1100007863.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100155968.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155968.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155968.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155968.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155968.hg.1 | TC1100007863.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100155971.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155971.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155971.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155971.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155971.hg.1 | ENST00000525947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155971.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155971.hg.1 | TC1100007863.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100155972.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155972.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155972.hg.1 | ENST00000278833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155972.hg.1 | NM_000327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155972.hg.1 | TC1100007863.hg.1 | Clariom D TC |

ROM1
Genomic Map