Assay Details
| Gene Symbol: NDUFA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018838.4 | NP_061326.1 | 2 - 3 | 205 | 145 | |
| GenBank mRNA | AF112208.1 | - | 2 - 3 | 225 | 145 | |
| AF217092.1 | - | 2 - 3 | 184 | 145 | ||
| AK222619.1 | - | 2 - 3 | 192 | 148 | ||
| BC005936.1 | - | 2 - 3 | 194 | 4069306 | 145 | |
| BG943528.1 | - | 2 - 3 | 190 | 145 | ||
| BM548072.1 | - | 272 | 145 | |||
| BT007220.1 | - | 2 - 3 | 172 | 145 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001827.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12024070.hg.1 | ENST00000327772.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024070.hg.1 | ENST00000547986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024070.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024070.hg.1 | TC12001827.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009625.hg.1 | ENST00000455914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000547157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000547157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000547157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024072.hg.1 | ENST00000327772.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024072.hg.1 | ENST00000547157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024072.hg.1 | ENST00000547986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024072.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024072.hg.1 | TC12001827.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024068.hg.1 | ENST00000327772.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024068.hg.1 | ENST00000550187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024068.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024068.hg.1 | TC12001827.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010633.hg.1 | ENST00000511389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204147.hg.1 | TC1200012829.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000538372.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000547157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000547157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000547157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000550187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012829.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | ENST00000547157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | ENST00000547157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204154.hg.1 | TC1200012829.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | ENST00000327772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | ENST00000546788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | ENST00000547986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | ENST00000551991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | NM_001258338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | NM_018838.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204151.hg.1 | TC1200012829.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010633.hg.1 | ENST00000511389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009625.hg.1 | ENST00000455914.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 174805 | - | - |

NDUFA12
Genomic Map