Assay Details
| Gene Symbol: CCRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130910.1 | NP_001124382.1 | 1 - 2 | 136 | 68 | |
| GenBank mRNA | AF015524.1 | - | 1 - 2 | 70 | 68 | |
| AK296673.1 | - | 1 - 2 | 136 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000257.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF015524.1,AK296673.1,NM_001130910.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03005221.hg.1 | TC03000257.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005221.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005221.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005221.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005221.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005230.hg.1 | TC03000257.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005230.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005230.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005230.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005230.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005230.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005230.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000433848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000433848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000441909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000441909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005223.hg.1 | TC03000257.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005223.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005223.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005223.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000433848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000433848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000441909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000441909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151442.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151442.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151442.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151442.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151442.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151442.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151442.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151442.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151442.hg.1 | TC0300007259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | ENST00000433848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | ENST00000433848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151452.hg.1 | TC0300007259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300151441.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151441.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151441.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151441.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151441.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151441.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151441.hg.1 | TC0300007259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300151444.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151444.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151444.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151444.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151444.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151444.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151444.hg.1 | TC0300007259.hg.1 | Clariom D TC |

CCRL2
Genomic Map