Assay Details
| Gene Symbol: WDR20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242415.1 | NP_001229344.1 | 4 | 1051 | 57 | |
| XM_011537336.2 | XP_011535638.1 | 5 | 2330 | 57 | ||
| XM_017021770.1 | XP_016877259.1 | 4 | 2251 | 57 | ||
| XM_017021785.1 | XP_016877274.1 | 5 | 2277 | 57 | ||
| GenBank mRNA | AK054748.1 | - | 1 | 154 | 57 | |
| AK130824.1 | - | 1 | 737 | 57 | ||
| BC030654.2 | - | 4 | 1030 | 57 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

WDR20
Genomic Map